ContactUsLinkComponent
WaterPro® PS HPLC/UF Hybrid Polishing Station
WaterPro® PS HPLC/UF Hybrid Polishing Station
Catalog # LBCP9000732
Supplier:  LABCONCO
undefined
WaterPro® PS HPLC/UF Hybrid Polishing Station
Catalog # LBCP9000732
Supplier:  LABCONCO
Specifications
  • Description:
    Waterpro PS HPLC/UF hybrid with dispensing gun
  • Electrical:
    230 V (EU)
  • Pk:
    1
Specifications
About this item
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. Labconco has designed WaterPro PS Stations with the right combinations of purification technologies to meet your needs.
The WaterPro PS/UF Hybrid model includes four bowls constructed of polypropylene for one carbon, two deionization and one organic adsorbtion cartridge (all cartridges sold separately).