Lab Pack Poly Drums
Catalog # EAMF1601B
Supplier: EAGLE MFG
Specifications
- Capacity:113 L
- Colour:Blue
- Description:Lab pack open head plastic barrel drum, plastic lever-lock
- Dimensions:485 mm Top, 413 mm Bottom, 670 mm Height
- Pk:1
- Weight:7,25 kg
Specifications
About this item
These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.
- Chemical and weather resistant
- UV protection
- Approved for use with packing groups i, ii and iii
- Regular lid
- 1H2/X100/S, 1H2/Y180/S
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.