
2-Chloroethanesulphonic acid sodium salt hydrate ≥98%
Catalog # APOSOR95369-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Purity:98%
Specification Test Results
About this item
:
:184.57 g/mol
Melting Pt:292 °C
:GHS07
CAS Number:1089719-72-1
:15484-44-3
Formula:C₂H₄ClNaO₃S.H₂O
MDL Number:MFCD00007531