Bromate-Bromide concentrate for dilution to 0,05 mol/L Br2 (0.1 N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32074.600
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:210 mL
- Seq:2545
Specifications
Specification Test Results
- Concentration after dilution to 1000 ML, c(Br2):0.05 mol/l ± 0.2% (20 °C)
Specification Test Results
About this item
:
:Dose for preparation of 1 litre of solution.
:GHS08
Storage Temperature:Ambient
:7758-01-2
Formula:KBrO₃/KBr