About this item
Permanent autoclave-resistant labels withstand various sterilisation procedures, including in steam autoclaves, dry heat ovens, gamma irradiators, and ethylene oxide gas sterilisers.
- Temperature range: −60 to 150 °C
- Ideal for use in repeated steam autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and ethylene oxide sterilisation
- Printable with all major thermal transfer printer brands
- Waterproof and weatherproof
- RR-class resin ribbon recommended
High temperature-resistant permanent labels for identifying various sterilisation containers and surfaces that will undergo autoclaving and dry heat sterilisation. These labels can be applied to metal, plastic, and glass surfaces, and will withstand up to 20 autoclave sterilisation cycles at 121 °C and 16,5 psi. The labels and the printout can withstand high temperature exposure, as well as gamma irradiation (up to 50 KGy) and ethylene oxide sterilisation. Ideal for identifying containers and surfaces that need to be sterilised inside autoclaves, dry heat sterilising equipment, and with gamma irradiators.