Order Entry
Northern Ireland
ContactUsLinkComponent
Free PSA (Human) ELISA kit
Free PSA (Human) ELISA kit
Catalog # ABNOKA6710.1KIT
Supplier:  Abnova
CAS Number:  
Free PSA (Human) ELISA kit
Catalog # ABNOKA6710.1KIT
Supplier:  Abnova
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    Free PSA
  • Assay Principle:
    Quantitative

Specifications

About this item

Free PSA (Human) ELISA Kit is an enzyme immunoassay for the quantitative measurement of free prostate-specific antigen in serum and plasma.