Anti-Paxillin-LD4 motif (residues 260-278) fused to MBP Rabbit Recombinant Antibody [clone: LD4 SAB]
Catalog # ABNORAB03733.200UG
Supplier: Abnova
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen symbol:Paxillin-LD4 motif (residues 260-278) fused to MBP
- Clonality:Recombinant
- Clone:LD4 SAB
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP.
- Pk:200 µG