Order Entry
Northern Ireland
ContactUsLinkComponent
Dispenser for hand disinfectants and lotions, plastic, sm2
Dispenser for hand disinfectants and lotions, plastic, sm2
Catalog # SCUE668500
Supplier:  SCHUELKE MAYR
undefined
Dispenser for hand disinfectants and lotions, plastic, sm2
Catalog # SCUE668500
Supplier:  SCHUELKE MAYR

Specifications

  • Colour:
    White
  • Description:
    Dispenser for 500/1000 ml bottles, sm2
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

Wall-mountable plastic dispenser for use with hand disinfectants and lotions (500 and 1000 ml bottles).

  • No contamination and evaporation thanks to the unique pump design and integrated bottle cap
  • Valve protected against contact and contamination
  • Almost complete emptying of the container thanks to the flexible suction hose
  • Adjustable dosage stroke: 1, 2 or 3 ml
  • Easy cleaning and, if necessary, disinfection thanks to easy dismantling and complete dismantling of the dispenser and pump
  • Easy mounting