Dispensers for hand disinfectants and lotions, metal, KHL / KHK
Catalog # SCUE669600
Supplier: SCHUELKE MAYR
Specifications
- Description:Dispenser KHK, for 500/1000 ml bottles, approximately 0,75 - 1,5 ml per stroke
- Seq:02
- Pk:1
Specifications
About this item
Wall-mountable dispensers for use with hand disinfectants and lotions (500 and 1000 ml bottles).
- Adjustable dosage stroke from approx. 0,75 ml to approx. 1,5 ml
- Housing made from aluminum, anodized (has a bacteriostatic effect)
- Dispenser is dishwasher safe
- Type KHL is autoclavable
- Easy wall mounting