Transfection reagent, 293-Free™
Catalog # 72181-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Pk:1 mL
- Cell specific:293
- DNA:Yes
- Description:Novagen® 293-Free™ transfection reagent
Specifications
About this item
293-Free™ transfection reagent has a unique polycationic liposomal formulation and is designed expressly for transfection of HEK 293 cells grown in suspension culture. It is ideal for mammalian protein production. 293-Free™ transfection reagent is provided as a sterile, ready to use solution. The 1 ml size provides enough reagent to transfect a litre of culture.
- Optimised for transient transfections of HEK 293 suspension cultures
- Derived from non-animal sources
- Minimal cellular toxicity
- Protocol easily scales up for production
- Compatible with both serum-containing and serum-free media
- Eliminates the need for media changes