Order Entry
Northern Ireland
ContactUsLinkComponent
Selenium reagent mixture for the determination of nitrogen acc. to Wieninger
Irritant
Selenium reagent mixture for the determination of nitrogen acc. to Wieninger
Catalog # 92504.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Selenium reagent mixture for the determination of nitrogen acc. to Wieninger
Catalog # 92504.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Na2SO4 (sodium sulfate):
    97.00%
  • CuSO4 (copper sulfate):
    1.50%
  • Se (selenium):
    1.50%
  • Application test (total nitrogen):
    conforms

Specification Test Results

About this item

 :
 :GHS07
Storage Temperature:Ambient