Safety Boots, Slip-On, High, Classic Safety Wellington, S4
Catalog # PRTWFW94BKR48
Supplier: Portwest
Specifications
- Shoe size:48
- Colour:Black
- Pk:1 PAIR
Specifications
About this item
Safety wellington is manufactured from virgin PVC/Nitrile in a seamless watertight construction. Ideal for use in a wide range of environments and industries.
- Reinforced ankle protection
- Chemical-resistant
- Protective steel toecap
- Anti-static footwear
- Energy absorbing seat region
- 100% waterproof to keep feet dry
- SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
- Fuel- and oil-resistant outsole
Classic safety wellington features include a 200 joule steel toecap, washable nylon lining, and kick off spurs.
Certifications: EN ISO 20345: 2011 S4. EN 13832-3: 2018. CE certified.