The SHARPSGUARD® eco yellow range is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic or cytostatic.
- Sustainable alternative made from at least 40% recycled plastic
- Small models can be easily fitted into a bag or glove compartment
- Models eco yellow 7 and above are supplied with a robust pail handle for easy transportation
These containers are for disposal by incineration or other authorised process and designed for the consignment of waste categorised by the following EWC codes: 18 01 01, 18 01 03*, 18 01 09, 18 02 01, 18 02 02*, 18 02 08.