Indoor/Outdoor Vinyl Labels for M710
Supplier: Brady
Certificates
About this item
B-595 vinyl will adhere to difficult low surface energy items, including PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics. The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
- Permanent acrylic adhesive keeps labels adhered in harsh climates and is resistant to chemicals, oils and detergents
- Vinyl tape is ideal in both indoor and outdoor use
- Adheres to surfaces that are smooth, rough or powder coated, and to irregular surfaces or surfaces with low surface energy
- Prints on white vinyl background with a clean gloss finish that is resistant to exposure to chemicals, grease, oil, and cleaning agents
- Average outdoor durability of 8-10 years in temperatures ranging from −40 to 82 °C (−40 to +180 °F)
- Gloss finish
Ideal for component and equipment labelling, electrical labelling, facility identification, floor marking, laboratory labelling, lean and 5S labelling, pipe and valve marking, and warehouse marking applications.
A ribbon is required to print this material. Use part number M7-R10000.
Certifications: BS5609:1986 section 2 and section 3.
Packaging: 1 roll per box.
Frequently Bought Together