ContactUsLinkComponent
Scotch-Weld™ DP190 Epoxy Adhesive Grey
Scotch-Weld™ DP190 Epoxy Adhesive Grey
Catalog # MMMA190.50
Supplier:  3M
undefined
Scotch-Weld™ DP190 Epoxy Adhesive Grey
Catalog # MMMA190.50
Supplier:  3M
Specifications
  • Volume:
    48,5 ml
  • Colour:
    Grey
  • Description:
    Scotch-Weld™ Epoxy adhesive, DP190
  • Pk:
    12
Specifications
About this item
Scotch-Weld™ Epoxy adhesive DP190 is a two-part epoxy adhesive that delivers exceptional performance with high shear and peel strength. Extremely compatible, it is formulated to bond to a wide range of materials, including metals, ceramics, wood, fibreboard, glass, rubber, and many plastics.
  • Apply with ease with a low viscosity formulation
  • Two-part epoxy adhesive is highly compatible with many different materials
  • Formulated with low viscosity for easy dispensing
Scotch-Weld™ Epoxy Adhesive DP190 adheres with tough, strong bonds and offers an extended work life. The adhesive is designed with a low viscosity formulation to give additional time for adjustment and make application easy.