ContactUsLinkComponent
Fiberlite™ Rotor Adapters
Catalog # HERA75100378
Supplier:  Thermo Scientific
undefined
Fiberlite™ Rotor Adapters
Catalog # HERA75100378
Supplier:  Thermo Scientific
Specifications
  • Description:
    Rotor adapter
  • Pk:
    2
  • For:
    15 ml conical tubes
Specifications
About this item
Incorporate Thermo Scientific™ Fiberlite™ Rotor Adapters with Fiberlite F13-14x50c and F15-8x50c rotors.