Order Entry
Northern Ireland
ContactUsLinkComponent
Poloxamer 182 (Pluronic® L-62, Synperonic® L62)
Poloxamer 182 (Pluronic® L-62, Synperonic® L62)
Catalog # SPCMP1813-500ML
Supplier:  Spectrum Chemical
CAS Number:  
Poloxamer 182 (Pluronic® L-62, Synperonic® L62)
Catalog # SPCMP1813-500ML
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Cloud Point (10 % Aqueous):
    22 - 26 °C
  • pH of a 2.5% Aqueous Solution:
    5.0- 7.5
  • Color (APHA):
    max. 50
  • Water:
    max. 0.4 %

Specification Test Results

About this item

 :
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:MFCD00082049