Potassium-L-aspartate dihydrate
Catalog # SPCMA3204-50KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
About this item
:
:207.223 g/mol
Storage Temperature:Ambient
CAS Number:1115-63-5
Formula:C₄H₆NO₄·K·2H₂O
MDL Number:MFCD00021083