
di-Potassium oxalate monohydrate ≥98.5%, crystals ACS
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:6487-48-5
:
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033
:184.23 g/mol
Merck Index:12,07820
Melting Pt:356 °C
Density:2,154 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-506-8
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Assay:98.5% Min.
Specification Test Results
Frequently Bought Together