Order Entry
Northern Ireland
ContactUsLinkComponent
Calcium thioglycolate trihydrate
Irritant
Calcium thioglycolate trihydrate
Catalog # SPCMC1701-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Calcium thioglycolate trihydrate
Catalog # SPCMC1701-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
 :276.35 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:65208-41-5
 :MFCD00043437
Formula:C₄H₆CaO₄S₂·3H₂O
MDL Number:MFCD00150707