Dodecyltrimethylammonium bromide
10 kg
Catalog # SPCMD3009-10KGBL
Supplier: Spectrum Chemical
Dodecyltrimethylammonium bromide
Catalog # SPCMD3009-10KGBL
Supplier: Spectrum Chemical
CAS Number: 1119-94-4
Jump to:
Product Details & Documents
Dodecyltrimethylammonium Bromide is a quaternary ammonium salt useful as paint strippers, foaming stabilizers, and bactericidal lotions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonyms: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide
- CAS Number:1119-94-4
- Formula:C₁₅H₃₄BrN
- MDL Number:MFCD00011767
- Molecular Weight:308.35 g/mol
- Melting Pt:264…266 °C
- Storage Temperature:Ambient
- EINECS:214-290-3
Specifications
Specifications
- Pk:10 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



