Oxybenzone ≥97% USP
Catalog # SPCMB1261-25GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Glass, amber
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Assay:97% Min.
Specification Test Results
About this item
:
:228.25 g/mol
Merck Index:12,07088
Boiling Pt:370 °C (1013 hPa)
Melting Pt:65 °C
Density:1,3397 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:205-031-5
CAS Number:131-57-7
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00008387