Potassium chloride ≥99%, granular USP
Catalog # SPCMPO170-500GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
About this item
Synonyms: Potassiumchloride
:
:74.55 g/mol
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1,98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360
Merck Index:14,07621