Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:C20orf165
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:21-120/227
- Form:Liquid
- Gene ID:128497
- Antigen synonyms:Uncharacterized protein C20orf165|C20orf165|TSG23|dJ337O18.8|CT165_HUMAN|Chromosome 20 open reading frame 165
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:24 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C20orf165
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf165 gene product has been provisionally designated C20orf165 pending further characterization.
Type: Primary
Antigen: C20orf165
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 21-120/227
Host: Rabbit
Isotype: IgG
Reactivity: