Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:C1orf43
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:11-100/253
- Form:Liquid
- Gene ID:25912
- Antigen synonyms:4933434E20Rik|Protein NICE 3|Protein NICE-3|AI462154|HSPC012|MGC111001|C1orf43|Riken cDNA 4933434E20|S863 3|OTTHUMP00000034199|Hepatitis C virus NS5A-transactivated protein 4|HCV NS5A transactivated protein 4|S863-3|OTTHUMP00000034201|NICE3|OTTHUMP00000034202|Chromosome 1 open reading frame 43|Hepatitis C virus NS5A transactivated protein 4|Hypothetical protein LOC25912|NS5ATP4|HCV NS5A-transactivated protein 4|Uncharacterized protein C1orf43|CA043_HUMAN|NICE 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:29 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C1orf43
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C1orf43, also known as Hepatitis C virus NS5A-transactivated protein 4 and Protein NICE-3, is a 253 amino acid single-pass membrane protein. There are five isoforms of C1orf43 that are produced as a result of alternative splicing events. The gene encoding C1orf43 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1.
Type: Primary
Antigen: C1orf43
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 11-100/253
Host: Rabbit
Isotype: IgG
Reactivity: Human