Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:LAP2 alpha
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/694
- Form:Liquid
- Antigen synonyms:TP|Thymopoietin isoform alpha|Thymopoietin|TP alpha|Isoforms beta/gamma|LAP2A_HUMAN|Lamina associated polypeptide 2|LAP2|LAP2 alpha|TMPO|Lamina associated polypeptide 2 alpha
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:83 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human LAP2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The nuclear envelope separates the nucleoplasm from the cytoplasm in eukaryotic cells and includes the outer and inner nuclear membrane, nuclear pore complexes and the nuclear lamina. The nuclear lamina contains intermediate filament-type proteins called lamins that form a dense network to strengthen and stabilize the nuclear envelope. Lamina-associated polypeptide 2 (LAP2) is also known as thymopoietin. LAP2 is a nuclear envelope protein and contains an amino-terminal region called the LAP2-emerin-MAN1 or LEM motif. LAP2 also contains a unique DNA-binding amino-terminal domain. Alternative splicing produces six isoforms (, _,_, and ) of mammalian LAP2 and three isoforms in Xenopus LAP2. LAP2 and LAP2_ associate with chromosomal barrier-to-autointegration factor (BAF) and may play a role in stabilizing chromatin structure. LAP2_ also binds to lamin B. LAP2 is a non-membrane isoform of LAP2 that associates with the internal nucleoskeleton and binds lamin A. The gene encoding human LAP2 maps to chromosome 12q23.1.
Type: Primary
Antigen: LAP2 alpha
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-150/694
Host: Rabbit
Isotype: IgG
Reactivity: