Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:BAIAP2L2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/529
- Form:Liquid
- Antigen synonyms:FLJ22582|BAI1-associated protein 2-like protein 2|BAI1-associated protein 2-like 2|BAIAP2L2 BAI1-associated protein 2-like 2|baiap2l2|Planar intestinal- and kidney-specic BAR domain protein|RP5-1039K5.3|BI2L2_HUMAN|Brain-specic angiogenesis inhibitor 1-associated protein 2-like protein 2|Pinkbar|BC048937
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:59 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BAIAP2L2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The Src homology 3 (SH3) domain is a highly conserved 60 amino acid protein domain that is organized into a beta-barrel fold consisting of five or six beta strands arranged as two tightly packed anti-parallel beta sheets. This domain is found in proteins that mediate assembly of specific protein complexes and interact with other proteins, specifically recognizing proline-rich regions. BAIAP2L2 (Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2) is a 529 amino acid protein containing an SH3 domain, suggesting that it may function as an adaptor protein. BAIAP2L2 also contains an IMD (IRSp53/MIM) domain, which enables the protein to bind to and bundle Actin filaments, as well as bind to membranes and interact with Rac GTPase. There are two named isoforms of BAIAP2L2 which are produced as a result of alternative splicing events.
Type: Primary
Antigen: BAIAP2L2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/529
Host: Rabbit
Isotype: IgG
Reactivity: