Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Gemin 7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Epitope:21-120/131
- Form:Liquid
- Gene ID:79760
- Antigen synonyms:Gemin7|GEMI7_HUMAN|SIP3|Gemin-7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:15 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Gemin 7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Gemin7 is a 131 amino acid protein encoded by the mouse gene Gemin7. Gemin7, along with Gemin6, is a significant component of the the large multiprotein human SMN complex. The SMN complex functions as an assembly machine for small nuclear ribonucleoproteins (snRNPs)-the major components of the spliceosome. The survival of motor neurons (SMN) protein, a product of the disease gene of the common neurodegenerative disease, spinal muscular atrophy, is also part of the SMN complex. Although Gemin6 and Gemin7 have no significant sequence similarity with Sm proteins, both adopt canonical Sm folds. Moreover, Gemin6 and Gemin7 exist as a heterodimer, and interact with each other via an interface similar to that which mediates interactions among the Sm proteins. The Gemin6/Gemin7 complex binds to Sm proteins and might help organize Sm proteins for formation of Sm rings on snRNA targets.
Type: Primary
Antigen: Gemin 7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/131
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat