Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:UBE2D2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:31-130/147
- Form:Liquid
- Gene ID:7322
- Antigen synonyms:Ubiquitin conjugating enzyme E2D 2 homologous to yeast UBC4/5|UB2D2_HUMAN|E217KB2|UBCH 5B|PUBC 1|Ubiquitin-conjugating enzyme E217KB 2|UBC 4|UBC4/5 homolog yeast|UBCH5B|Ubiquitin conjugating enzyme E2 D2|PUBC1|Ubiquitin carrier protein|Ubiquitin carrier protein D2|UBC 4/5|UBC4/5|Ubiquitin-conjugating enzyme E2-17 kDa 2|Ubiquitin conjugating enzyme E2 17 kDa 2|Ubiquitin-conjugating enzyme E2 D2|Ubiquitin conjugating enzyme E2D 2 UBC4/5 homolog yeast|Ubiquitin-protein ligase D2|Ubiquitin conjugating enzyme E2D 2|UBC4|Ubiquitin protein ligase D2|UBE2D2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:17 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human UBE2D2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Ubiquitin is an abundant, highly conserved protein found in all eukaryotic cells either free or covalently attached to cellular proteins. The primary function of ubiquitin in mammalian systems is to clear abnormal, foreign, and improperly folded proteins by targeting them for proteosome degradation. UBE2D proteins, including UBE2D1 (ubiquitin-conjugating enzyme E2D1 or UBC5A), UBE2D2 (ubiquitin-conjugating enzyme E2D2 or UBC5B) and UBE2D3 (ubiquitin-conjugating enzyme E2D3 or UBC5C), are E2 ubiquitin-conjugating enzymes that catalyze the ubiquitination of I˚Bå in a phosphorylation and SCFB-TRCP-dependent manner. Specifically, E1 first transfers a ubiquitin residue to the E2 component (a UBE2D protein), and the UBE2D protein then associates with an E3 ubiquitin-protein ligase, which immediately transfers that residue to a protein that is targeted for degradation.
Type: Primary
Antigen: UBE2D2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-130/147
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse