Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:CstF-64
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:61-160/616
- Form:Liquid
- Gene ID:23283
- Antigen synonyms:CF1 64 kDa subunit, tau variant|Cleavage stimulation factor 64 kDa subunit, tau variant|TauCstF64|KIAA0689|CSTF 64 kDa subunit, tau variant|CSTFT_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:64 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CSTF2T/CstF-64
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Polyadenylation of mRNA precursors is a two-step reaction that requires multiple protein factors. The first step, endonucleolytic cleavage of polyadenylation substrates, requires CstF (cleavage stimulation factor), a heterotrimer that is composed of three distinct subunits. CstF-64 contains an RNA binding domain and is responsible for the RNA binding activity of CstF. CstF-64 is expressed in all somatic cells and in pre- and postmeiotic, but not meiotic, germ cells. However, a large variant of CstF-64, called t CstF-64, is abundantly expressed in meiotic and postmeiotic cells in the testis and to a lesser extent in the brain, and promotes the germ cell pattern of polyadenylation. The gene encoding CstF-64 (designated CSTF2) maps to the X chromosome, whereas t CstF-64 is encoded by an autosomal gene. The increase in CstF-64 concentration during B cell activation switches IgM heavy chain mRNA expression from membrane-bound to secreted forms, suggesting that CstF-64 plays a key role in regulating IgM heavy chain expression during B cell differentiation.
Type: Primary
Antigen: CstF-64
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 61-160/616
Host: Rabbit
Isotype: IgG
Reactivity: