Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:ALDH9A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:401-494/494
- Form:Liquid
- Gene ID:223
- Antigen synonyms:R-aminobutyraldehyde dehydrogenase|ALDH9|Aldehyde dehydrogenase 9A1|ALDH4|Aldehyde dehydrogenase family 9 member A1|AL9A1_HUMAN|4-trimethylaminobutyraldehyde dehydrogenase|ALDH7|Aldehyde dehydrogenase E3 isozyme|Gamma aminobutyraldehyde dehydrogenase|R aminobutyraldehyde dehydrogenase|E3|TMABADH|4 trimethylaminobutyraldehyde dehydrogenase|Gamma-aminobutyraldehyde dehydrogenase|aldehyde dehydrogenase NAD+|aldh9A1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:54 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ALDH9A1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH9A1 (aldehyde dehydrogenase family 9 member A1), also known as E3, ALDH4, ALDH7, ALDH9 or TMABADH, is a 494 amino acid cytoplasmic protein that is highly expressed in adult liver, skeletal muscle, kidney and embryonic brain. ALDH9A1 converts gamma-trimethylaminobutyraldehyde into gamma-butyrobetaine and catalyses the irreversible oxidation of a broad range of aldehydes to the corresponding acids in a NAD-dependent reaction.
Type: Primary
Antigen: ALDH9A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-494/494
Host: Rabbit
Isotype: IgG
Reactivity: