Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:RPGRIP1L
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:41-140/1315
- Form:Liquid
- Antigen synonyms:Protein fantom|CORS 3|Retinitis pigmentosa GTPase regulator interacting protein 1 like|NPHP 8|NPHP8|Meckel syndrome, type 5|JBTS1|JBTS 1|Fantom|JBTS 7|MKS 5|MKS5|FTM_HUMAN|RPGRIP1-like|JBTS7|Meckel syndrome, type 1|RPGR interacting protein 1 like protein|Joubert syndrome 1|CORS3|nephrocystin 8|Joubert syndrome 7|FTM|RPGRIP1 like protein
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:151 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RPGRIP1L
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
RPGRIP1L is a 1315 amino acid protein that belongs to the RPGRIP1 family and is thought to function in programmed cell death, craniofacial development and formation of the left-right axis. Existing as two alternatively spliced isoforms that localize to the cytoplasm, cytoskeleton, centrosome and cilium basal body, RPGRIP1L interacts with nephrocystin-4 and is moderately expressed in brain, retina and kidney. Containing two C2 domains, RPGRIP1L is encoded by a gene that maps to human chromosome 16q12.2. Defects in the gene encoding RPGRIP1L are the cause of Joubert syndrome type 7 (JBTS7), COACH syndrome (COACHS) and Meckel syndrome type 5 (MKS5).
Type: Primary
Antigen: RPGRIP1L
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-140/1315
Host: Rabbit
Isotype: IgG
Reactivity: