Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CSDC2/PIPPIN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:21-120/153
- Form:Liquid
- Antigen synonyms:RNA binding protein PIPPin|Csdc2|Cold shock domain containing C2 RNA binding|CSDC2_HUMAN|Cold shock domain-containing protein C2|Cold shock domain containing protein C2|dJ347H13.2|PIPPIN|RNA-binding protein PIPPin|CSDC 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:17 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PIPPIN
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
PIPPIN is a Y-box protein (also called cold-shock (CSD) domain-containing protein) and belongs to a family of highly conserved RNA-binding transcriptional regulators. Predominantly expressed in brain cells and localizing to the nucleus and the cytoplasm, PIPPIN contains two RNA-binding motifs, namely PIP1 and PIP2, and one CSD domain. PIPPIN functions as a nucleic acid binding regulatory factor and is believed to participate in brain maturation. More specifically, PIPPIN binds to the 3 UTR ends of the mRNAs encoding Histone H1 and Histone H3.3. This interaction requires all of the PIPPIN domains to work in concert as one functional protein. In addition, PIPPIN can be sumoylated in a thyroid hormone (T3)-dependent manner. This suggests that PIPPIN modification in response to extracellular stimuli may modulate the regulation of protein synthesis.
Type: Primary
Antigen: CSDC2/PIPPIN
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/153
Host: Rabbit
Isotype: IgG
Reactivity: