Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-alpha + beta Synuclein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-alpha + beta Synuclein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11787R-A750
Supplier:  Bioss
Anti-alpha + beta Synuclein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11787R-A750
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    alpha + beta Synuclein
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish,
    Human,
    Rat,
    Mouse
  • Epitope:
    32-44/140
  • Form:
    Liquid
  • Antigen synonyms:
    PARK1|SNCB|SYUB_HUMAN|SNCB protein|Parkinson disease familial 1|SNCA|PARK4|Beta synuclein|SYUA_HUMAN|Alpha synuclein|Non A4 component of amyloid precursor|Non A beta component of AD amyloid|NACP|PD1|Synuclein alpha|Synuclein beta
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    14 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human alpha+beta Synuclein
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Synucleins are small soluble proteins expressed primarily in neural tissues and in certain tumours. The family includes 3 known proteins, alpha synuclein, beta synuclein and gamma synuclein. All synucleins have in common a highly conserved alpha helical lipid binding motif with similarity to the class A2 lipid binding domains of the exchangeable apolipoproteins. The alpha and beta synuclein proteins are found primarily in brain tissue, where they are seen mainly in pre synaptic terminals. Alpha synuclein is believed to be a major component of Lewy bodies in Parkinson's disease. Mutations in alpha synuclein are associated with rare familial cases of early onset Parkinson's disease, Alzheimer's disease and several other neurodegenerative illnesses.

Type: Primary
Antigen: alpha + beta Synuclein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 32-44/140
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish