Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:OR5T1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:88-111/326
- Form:Liquid
- Antigen synonyms:olfactory receptor, family 5, subfamily T, member 1|Olfactory receptor 5T1| OR5T1_HUMAN|Olfactory receptor OR11-179|OR5T1P|OR11-179
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human OR5T1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
OR5T1.
Type: Primary
Antigen: OR5T1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 88-111/326
Host: Rabbit
Isotype: IgG
Reactivity: