Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ET-2/Endothelin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-ET-2/Endothelin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11280R-A680
Supplier:  Bioss
Anti-ET-2/Endothelin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11280R-A680
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ET-2/Endothelin 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Cat
  • Epitope:
    49-59/178
  • Form:
    Liquid
  • Antigen synonyms:
    EDN2|ET 2|ET-2|Endothelin 2|Preproendothelin-2|EDN2_HUMAN|Endothelin-2|PPET2|ET2|Preproendothelin 2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    2,3 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Endolin 2
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The endothelin (ET) family of proteins, which includes ET-1 (endothelin-1), ET-2 (endothelin-2) and ET-3 (endothelin-3), are vasoactive peptides that are involved in various functions throughout the body. Endothelins can affect the central nervous system and neuronal excitability, and they elicit potent vasoconstrictor action. While ET-1 is a potent, 21-amino acid vasoconstrictor peptide, ET-2 has the most potent vasoconstrictor activity. ET-3 functions as a ligand for endothelin receptor type B (ETBR) and, through this interaction, mediates the maturation of enteric neurons and melanocytes. Although ET-3 is expressed as a 238 amino acid peptide, it is post-translationally modified to produce a short active isoform and a long inactive isoform. Defects in the gene encoding ET-3 are the cause of a variety of disorders, including Hirschsprung disease type 1 (HSCR1), congenital central hypoventilation syndrome (CCHS) and Waardenburg syndrome type IV (WS4).

Type: Primary
Antigen: ET-2/Endothelin 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 49-59/178
Host: Rabbit
Isotype: IgG
Reactivity: Cat