Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Cyclophilin 40
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:281-370/370
- Form:Liquid
- Gene ID:5481
- Antigen synonyms:40 kDa peptidyl prolyl cis trans isomerase D|MGC33096|CYP 40|PPID|Rotamase D|Peptidyl-prolyl cis-trans isomerase D|CYPD|CyclophilinD|40 kDa peptidyl-prolyl cis-trans isomerase|Cyclophilin-40|Rotamase|Cyclophilin related protein|PPID_HUMAN|Cyclophilin D|CYP-40|40 kDa peptidyl prolyl cis trans isomerase|PPIase D|Peptidylprolyl isomerase D|CYP40|Cyclophilin-related protein|PPIase|Cyclophilin40|Peptidyl Prolyl Isomerase D
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Cyclophilin 40
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Cyclophilins are conserved, ubiquitous and abundant cytosolic peptidyl-prolyl cis-trans isomerases that accelerate the isomerization of XaaPro peptide bonds and the refolding of proteins. Human cyclophilin A (CyPA), an intracellular protein of 165 amino acids, is the target of cyclosporin A (CsA) and is encoded by a single unique gene conserved from yeast to humans. Cyclophilin B (CyPB) is secreted in biological fluids such as blood or milk and binds to a specific receptor present on the human lymphoblastic cell line Jurkat and on human peripheral blood lymphocytes. Cyclophilin D (CyP40) is a widely expressed cytoplasmic protein that catalyses the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. It is a widely expressed cytoplasmic protein.
Type: Primary
Antigen: Cyclophilin 40
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 281-370/370
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat