Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-C20orf196 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-C20orf196 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9699R-A680
Supplier:  Bioss
Anti-C20orf196 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9699R-A680
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C20orf196
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    41-140/205
  • Form:
    Liquid
  • Gene ID:
    149840
  • Antigen synonyms:
    SHLD1|Shieldin complex subunit 1|SHLD1_HUMAN|C20orf196|Uncharacterized protein C20orf196|CT196_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C20orf196
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf196 gene product has been provisionally designated C20orf196 pending further characterisation.

Type: Primary
Antigen: C20orf196
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-140/205
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat