Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Ghrelin 28
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:24-51/117
- Form:Liquid
- Gene ID:58991
- Antigen synonyms:motilin-related peptide|grehlin|growth hormone secretagogue|protein M46|growth hormone-releasing peptide|Appetite-regulating hormone
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:13 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide of mouse Ghrelin
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release. Ghrelin is derived from a preprohormone called preproghrelin, which also generates a second peptide called obestatin. Obestatin is an endogenous ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and decreased food intake. Also known as Appetite regulating hormone; GHRL; Growth hormone releasing peptide; Growth hormone secretagogue; M46 protein; Motilin related peptide; MTLRP; Obestatin; Obestatin preprohormone; PRO1066; UNQ524. Sequence notes: Gly-Ser-Ser-Phe-Leu-Ser-Pro- Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu- Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro- Arg (mo, rat).
Type: Primary
Antigen: Ghrelin 28
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 24-51/117
Host: Rabbit
Isotype: IgG
Reactivity: Rat