Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Defensin alpha 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Defensin alpha 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2799R-A750
Supplier:  Bioss
Anti-Defensin alpha 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2799R-A750
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Defensin alpha 4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    41-97/97
  • Form:
    Liquid
  • Gene ID:
    1669
  • Antigen synonyms:
    Defensin, alpha 4|HP-4|neutrophil defensin 4 precursor|defensin, alpha 4, corticostatin|Neutrophil defensin 4|HNP-4|neutrophil defensin 4 preproprotein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    3.8 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Defensin alpha 4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defence. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes are clustered on chromosome 8. This gene differs from other genes of this family by an extra 83-base segment that is apparently the result of a recent duplication within the coding region. The protein encoded by this gene, defensin, alpha 4, is found in the neutrophils; it exhibits corticostatic activity and inhibits corticotropin stimulated corticosterone production.

Type: Primary
Antigen: Defensin alpha 4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-97/97
Host: Rabbit
Isotype: IgG
Reactivity: