Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:PPIL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:1-130/166
- Form:Liquid
- Gene ID:51645
- Antigen synonyms:PPIL1|Peptidylprolyl isomerase cyclophilin like 1|CGI124|hCyPX|CGI 124|cyclophilin-related gene 1|peptidylprolyl isomerase cyclophilinlike1|Cyclophilin related gene 1|Peptidyl-prolyl cis-trans isomerase-like 1|peptidylprolyl isomerase like 1|CGI-124|peptidylprolyl isomerase cyclophilinlike 1|CYPL1|PPIL1_HUMAN|cyclophilinrelated gene 1|peptidylprolyl isomerase cyclophilin-like 1|peptidyl-prolyl cis-trans isomerase|PPIase|Rotamase PPIL1|MGC678|rotamase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:18 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PPIL1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Peptidyl-prolyl cis-trans isomerase (PPIL1) is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyse the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
Type: Primary
Antigen: PPIL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-130/166
Host: Rabbit
Isotype: IgG
Reactivity: