Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Cadherin 20
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:301-400/801
- Form:Liquid
- Antigen synonyms:FLJ37047|CDH7L3|Cadherin 20, type 2|Cdh7|Cadherin 20|cadherin-20
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:82 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Cadherin 20
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterised by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. Cadherin-20, also known as protocadherin gamma B4, is a member of the protocadherin gamma gene cluster. Expressed primariy in fibroblasts, cadherin-20 plays a critical role in neuronal cell-cell connections in the brain. Through its ability to mediate cell-cell adhesion, caderin-20 is thought to help the body respond to injury by regulating post-traumatic wound healing.
Type: Primary
Antigen: Cadherin 20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/801
Host: Rabbit
Isotype: IgG
Reactivity: