Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:HCP5
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:1-80/132
- Form:Liquid
- Antigen synonyms:HCP5_HUMAN|HLA complex protein P5|HLA class I histocompatibility antigen protein P5|Protein P5 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HCP5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
HCP5 is a region present on chromosome 6p21.3 that is characterised by multiple duplicated gene families. HCP5 (HLA class I histocompatibility antigen protein P5), also known as P5-1, is a 132 amino acid protein that is encoded by a gene mapping to human chromosome 6p21.33. Localising within the MHC class I region, HCP5 is not structurally related to other MHC class I genes, but does have high sequence similarity with HERV-L and HERV-16. HCP5 also has high sequence homology to retroviral Pol genes, making it a possible candidate for interaction with HIV-1 through an antisense mechanism that prevents retrovirus transcription. Single-nucleotide polymorphisms (SNPs) to the region of the HCP5 gene that corresponds with HLA-B has been linked to a lower HIV-1 viral set point. HCP5 is highly expressed in lymphoid tissues, spleen and activated lymphocytes, as well as B-cell and natural killer (NK) cell lines.
Type: Primary
Antigen: HCP5
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 1-80/132
Host: Rabbit
Isotype: IgG
Reactivity: