Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:C9orf103
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/184
- Form:Liquid
- Gene ID:414328
- Antigen synonyms:OTTHUMP00000021547|Chromosome 9 open reading frame 103|GNTK_HUMAN|C9orf103|Gluconokinase like protein|Probable gluconokinase|Glucokinase like protein|OTTHUMP00000021546|Gluconate kinase|IDNK|RP11-522I20,2|bA522I20,2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:21 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C9ORF103
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C9orf103 (chromosome 9 open reading frame 103), also known as gluconate kinase, is a 187 amino acid protein that belongs to the gluconokinase gntK/gntV family and catalyses the conversion of ATP and D-gluconate to ADP and 6-D-gluconate. Existing as three alternatively spliced isoforms, the gene encoding C9orf103 maps to human chromosome 9q21.32. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster.
Type: Primary
Antigen: C9orf103
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-150/184
Host: Rabbit
Isotype: IgG
Reactivity: