Order Entry
Northern Ireland
ContactUsLinkComponent
 
COX Colorimetric Inhibitor Screening Assay Kit
Catalog # CAYM701050-192
Supplier:  Cayman Chemical
CAS Number:  
undefined
COX Colorimetric Inhibitor Screening Assay Kit
Catalog # CAYM701050-192
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    192 Tests
  • Description:
    Screening assay kit, buffer, heme, COX-1 (ovine), COX-2 (human), arachidonic acid (substrate), potassium hydroxide, substrate, plate, and cover sheet, 192 wells

Specifications

About this item

A colorimetric assay for the screening of isozyme-specific inhibitors.

  • Screen for inhibitors of ovine COX-1 or human COX-2
  • Measure the peroxidase component of the COX
  • Assay 41 samples for COX-1 and 41 samples for COX-2 in duplicate
  • Plate-based colorimetric measurement (590 nm)

Cayman's COX Colorimetric Inhibitor Screening Assay measures the peroxidase component of cyclooxygenases. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized N,N,N',N'-tetramethyl-p-phenylenediamine (TMPD) at 590 nm. Inhibition of COX activity, measured by TMPD oxidation, by a variety of selective and nonselective inhibitors, shows changed potencies similar to those observed with other in vitro methods. The COX Colorimetric Inhibitor Screening assay includes both ovine COX-1 and human recombinant COX-2 enzymes in order to screen isozyme-specific inhibitors. Cayman's COX Colorimetric Assay is a time saving tool for screening vast numbers of inhibitors.