Order Entry
Northern Ireland
ContactUsLinkComponent
PicoKine Rat Eotaxin; ELISA Kit
PicoKine Rat Eotaxin; ELISA Kit
Catalog # BSBTEK1435
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Rat Eotaxin; ELISA Kit
Catalog # BSBTEK1435
Supplier:  Boster Bio
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Target protein:
    Eotaxin (CCL11)
  • Description:
    Rat Eotaxin PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of rat Eotaxin.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect Rat Eotaxin/Ccl11 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Eotaxin/Ccl11 tissue specificity:

96wells/kit, with removable strips.