Order Entry
Northern Ireland
ContactUsLinkComponent
Klerwipe™ Sporicidal Enhanced Peroxide
Klerwipe™ Sporicidal Enhanced Peroxide
Catalog # SHIE6600533
Klerwipe™ Sporicidal Enhanced Peroxide
Catalog # SHIE6600533

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Packed:
    10 x 20 wipes
  • Description:
    Klerwipe Sporicidal Enhanced Peroxide Mop Wipe
  • L×W:
    420 x 250 mm
  • Pk:
    1

Specifications

About this item

Sterile low particulate mop wipes impregnated with an enhanced blend of 1.5% hydrogen peroxide. The biocide blend is 0.2 micron filtered then sterile filled and double bagged under Grade A laminar flow. A sterile ready-to-use safer, rotational sporicide mop wipe, in a peelable pack, for large surfaces.

Efficacity
Bactericide: EN 1276, EN 13697, EN 16615
Yeasticide: EN 13697, EN 1650, EN 16615
Fungicide: EN 13697, EN 1650
Virucide: EN 14476, RKI / DVV
Sporicide: EN 13697, EN 13704, EN 16615