Order Entry
Northern Ireland
ContactUsLinkComponent
 
Rhodium(III) chloride hydrate
Catalog # APOSIN3003-3-1G
Supplier:  Apollo Scientific
CAS Number:  
Rhodium(III) chloride hydrate
Catalog # APOSIN3003-3-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Rhodium trichloride hydrate
 :
 :227.28 g/mol
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:20765-98-4
Formula:Cl₃Rh·H₂O
MDL Number:MFCD00149839
Melting Pt:100 °C (dec.)