
3-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00645.9025
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Fibreboard box/case with liner
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Solubility:3 g/l (25 °C)
- Melting Point:139 - 142 °C
- Molar Mass:167.12 g/mol
- Bulk Density:700 kg/m3
- Flash Point:190 °C
- Density:1.49 g/cm3 (20 °C)
- pH:3 (5 g/l, H2O, 20 °C) (aqueous suspension)
- Ignition Point:490 °C
- Hill Formula:C7H5NO4
- Chemical Formula:3-(NO2)C6H4COOH
Specification Test Results
About this item
:
:167.12 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:121-92-6
Formula:C₇H₅NO₄
MDL Number:MFCD00007251