(±)-1,2-Propanediol ≥99.5%, meets analytical specification of BP, Ph. Eur., USP, Riedel-de Haën™
Catalog # HONC16033H-5L
Supplier: Honeywell Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Boiling range:184 - 189 °C
- Density (D 20/20):1.035 - 1.040
- Refractive index (n 20/D):1.431 - 1.433
- Water (Karl Fischer):Max. 0.2 %
- Sulfated ash:Max. 0.007 %
- Acidity:complying
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 70 ppm
- Sulfate (SO₄):Max. 60 ppm
- Oxidizing substances:complying
- Reducing impurities:complying
- Appearance of the substance:complying
- Residual solvents:complying
Specification Test Results
About this item
:
:76.1 g/mol
Boiling Pt:184 °C (1,003.2 hPa)
Melting Pt:-60 °C
Melting Pt:–60 °C
Density:1,03 g/cm3 (20 °C)
Flash Pt:103 °C
Storage Temperature:Ambient
CAS Number:57-55-6
Formula:CH₃CH(OH)CH₂OH
MDL Number:MFCD00064272